siRNA Details
| Gene Symbol: Muc20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145874.1 | NP_001139346.1 | 2 | 347 | ||
| NM_146071.2 | NP_666183.2 | 2 | 347 | |||
| GenBank mRNA | AK085640.1 | - | 2 | 314 | ||
| AK142548.1 | - | 2 | 348 | |||
| AK165477.1 | - | 2 | 339 | |||
| BB661244.1 | - | 2 | 348 | |||
| BC026367.1 | - | 2 | 348 | 4037422 | ||

Muc20
Genomic Map