siRNA Details
| Gene Symbol: 1700021K19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001200038.1 | NP_001186967.1 | 9 | 1550 | ||
| NM_172615.4 | NP_766203.1 | 8 | 1505 | |||
| GenBank mRNA | AK030368.1 | - | 8 | 1479 | ||
| AK042428.1 | - | 8 | 1443 | |||
| AK122221.1 | - | 9 | 1514 | |||
| AK136223.1 | - | 1 | 184 | |||
| AK170002.1 | - | 9 | 1443 | |||
| AK170223.1 | - | 9 | 1401 | |||
| AK170312.1 | - | 9 | 1694 | |||
| AK171583.1 | - | 8 | 1489 | |||
| BC057307.1 | - | 9 | 1454 | |||
| BC060601.1 | - | 9 | 1523 | 5706123 | ||
| BC067390.1 | - | 8 | 1490 | 30432960 | ||

1700021K19Rik
Genomic Map