siRNA Details
| Gene Symbol: Adcy5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012765.4 | NP_001012783.3 | 19 | 3952 | ||
| GenBank mRNA | AK147590.1 | - | 4055 | |||
| AK147649.1 | - | 4712 | ||||
| AK160942.1 | - | 19 | 3952 | |||
| BC035550.1 | - | 2090 | ||||
| BC090846.1 | - | 19 | 3687 | 30363147 | ||

Adcy5
Genomic Map