siRNA Details
| Gene Symbol: Dzip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110017.1 | NP_001103487.1 | 16 | 2309 | ||
| NM_027341.2 | NP_081617.1 | 15 | 1691 | |||
| GenBank mRNA | AK039172.1 | - | 16 | 2287 | ||
| BC052893.1 | - | 15 | 1499 | |||
| BC117953.1 | - | 15 | 1514 | 40098141 | ||
| BC117954.1 | - | 15 | 1520 | 40098143 | ||
| CF536693.1 | - | 3 | 712 | |||
| EU233997.1 | - | 8 | 1511 | |||

Dzip3
Genomic Map