siRNA Details
| Gene Symbol: Impg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174876.3 | NP_777365.2 | 11 | 1274 | ||
| GenBank mRNA | AY174061.1 | - | 11 | 1274 | ||
| BC048863.1 | - | 11 | 1243 | 5368568 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Impg2| Gene Symbol: Impg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174876.3 | NP_777365.2 | 11 | 1274 | ||
| GenBank mRNA | AY174061.1 | - | 11 | 1274 | ||
| BC048863.1 | - | 11 | 1243 | 5368568 | ||