siRNA Details
| Gene Symbol: Crybg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174848.3 | NP_777273.2 | 1 | 570 | ||
| GenBank mRNA | BC043118.1 | - | 1 | 570 | 6408859 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crybg3| Gene Symbol: Crybg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174848.3 | NP_777273.2 | 1 | 570 | ||
| GenBank mRNA | BC043118.1 | - | 1 | 570 | 6408859 | |