siRNA Details
Gene Symbol: Speer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173069.3 | NP_775092.3 | 4 | 461 | ||
GenBank mRNA | AY151401.1 | - | 3 | 232 | ||
BB610282.1 | - | 4 | 463 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Speer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173069.3 | NP_775092.3 | 4 | 461 | ||
GenBank mRNA | AY151401.1 | - | 3 | 232 | ||
BB610282.1 | - | 4 | 463 |