siRNA Details
| Gene Symbol: Scaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178923.3 | NP_849254.2 | 12 | 2082 | ||
| GenBank mRNA | AK045266.1 | - | 12 | 2067 | ||
| AK173120.1 | - | 12 | 2151 | |||
| BC023734.1 | - | 9 | 1483 | |||
| BC041782.1 | - | 9 | 1454 | |||
| BC053096.1 | - | 12 | 1965 | 6401143 | ||
| BC057592.1 | - | 12 | 2082 | 6417362 | ||

Scaf4
Genomic Map