siRNA Details
Gene Symbol: 4732491K20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029100.1 | - | 1, 2 | 288 | ||
CJ159212.1 | - | 1, 2 | 320 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4732491K20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029100.1 | - | 1, 2 | 288 | ||
CJ159212.1 | - | 1, 2 | 320 |