siRNA Details
| Gene Symbol: Vmn2r106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104568.1 | NP_001098038.1 | 1 | 71 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r106| Gene Symbol: Vmn2r106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104568.1 | NP_001098038.1 | 1 | 71 | ||