siRNA Details
| Gene Symbol: Zfp758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145484.2 | NP_663459.2 | 4 | 790 | ||
| GenBank mRNA | AK028301.1 | - | 3 | 674 | ||
| BC021442.1 | - | 4 | 687 | 5038660 | ||
| BC022203.1 | - | 4 | 767 | |||
| BC037455.1 | - | 4 | 791 | |||
| BC056632.1 | - | 3 | 648 | 5036441 | ||

Zfp758
Genomic Map