siRNA Details
| Gene Symbol: Rab40c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139154.2 | NP_631893.1 | 2, 3 | 251 | ||
| GenBank mRNA | AB232639.1 | - | 1, 2 | 145 | ||
| AF422144.1 | - | 1, 2 | 204 | |||
| BC027008.1 | - | 2, 3 | 239 | 5364768 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab40c| Gene Symbol: Rab40c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139154.2 | NP_631893.1 | 2, 3 | 251 | ||
| GenBank mRNA | AB232639.1 | - | 1, 2 | 145 | ||
| AF422144.1 | - | 1, 2 | 204 | |||
| BC027008.1 | - | 2, 3 | 239 | 5364768 | ||