siRNA Details
| Gene Symbol: Lemd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146075.2 | NP_666187.2 | 9 | 1761 | ||
| GenBank mRNA | AK082131.1 | - | 9 | 1569 | ||
| AK143224.1 | - | 9 | 1746 | |||
| BC026588.1 | - | 9 | 1071 | 4972900 | ||
| BC050010.1 | - | 7 | 1999 | |||

Lemd2
Genomic Map