siRNA Details
Gene Symbol: Zfp523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172617.3 | NP_766205.1 | 10 | 1136 | ||
GenBank mRNA | AK028222.1 | - | 10 | 1137 | ||
BC025615.1 | - | 7 | 701 | |||
BC058346.1 | - | 8 | 868 | 6832233 | ||
BC060611.1 | - | 9 | 1100 | 6416823 |
