siRNA Details
Gene Symbol: Zfp523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172617.3 | NP_766205.1 | 8, 9 | 962 | ||
GenBank mRNA | AK028222.1 | - | 8, 9 | 963 | ||
BC025615.1 | - | 5, 6 | 527 | |||
BC058346.1 | - | 6, 7 | 694 | 6832233 | ||
BC060611.1 | - | 7, 8 | 926 | 6416823 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
