siRNA Details
Gene Symbol: Zfp523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172617.3 | NP_766205.1 | 4 | 471 | ||
GenBank mRNA | AK028222.1 | - | 4 | 472 | ||
AK180200.1 | - | 2 | 139 | |||
BC058346.1 | - | 2 | 203 | 6832233 | ||
BC060611.1 | - | 3 | 435 | 6416823 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
