siRNA Details
| Gene Symbol: Zfp472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153063.3 | NP_694703.2 | 3 | 261 | ||
| GenBank mRNA | AK142395.1 | - | 3 | 252 | ||
| AY195875.1 | - | 3 | 196 | |||
| BB652931.1 | - | 3 | 262 | |||
| BC023090.1 | - | 3 | 196 | |||
| BC027407.1 | - | 3 | 251 | 4945483 | ||

Zfp472
Genomic Map