siRNA Details
| Gene Symbol: Tmem63b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198167.3 | NP_937810.2 | 19 | 1858 | ||
| GenBank mRNA | AK090200.1 | - | 19 | 1779 | ||
| AK159673.1 | - | 19 | 1863 | |||
| BC026370.1 | - | 2 | 117 | |||
| BC039277.1 | - | 8 | 840 | |||
| BC057136.1 | - | 8 | 821 | 6529222 | ||
| BC059283.1 | - | 12 | 1206 | |||

Tmem63b
Genomic Map