siRNA Details
| Gene Symbol: Tmem63b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198167.3 | NP_937810.2 | 14 | 1265 | ||
| GenBank mRNA | AK090200.1 | - | 14 | 1186 | ||
| AK159673.1 | - | 14 | 1270 | |||
| BC039277.1 | - | 3 | 247 | |||
| BC057136.1 | - | 3 | 228 | 6529222 | ||
| BC059283.1 | - | 7 | 613 | |||

Tmem63b
Genomic Map