siRNA Details
| Gene Symbol: Rrp36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144857.1 | NP_659106.1 | 4 | 321 | ||
| GenBank mRNA | AK148164.1 | - | 467 | |||
| AK148237.1 | - | 4 | 418 | |||
| BC011248.1 | - | 4 | 321 | 4192711 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rrp36| Gene Symbol: Rrp36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144857.1 | NP_659106.1 | 4 | 321 | ||
| GenBank mRNA | AK148164.1 | - | 467 | |||
| AK148237.1 | - | 4 | 418 | |||
| BC011248.1 | - | 4 | 321 | 4192711 | ||