siRNA Details
| Gene Symbol: Pex6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145488.1 | NP_663463.1 | 8 | 1803 | ||
| GenBank mRNA | AK143173.1 | - | 8 | 1791 | ||
| AK184357.1 | - | 2 | 121 | |||
| AK184585.1 | - | 2 | 120 | |||
| AK192148.1 | - | 2 | 121 | |||
| AK202266.1 | - | 2 | 122 | |||
| AK220525.1 | - | 8 | 1784 | |||
| BC003424.1 | - | 8 | 1803 | 2609884 | ||

Pex6
Genomic Map