siRNA Details
| Gene Symbol: Pex6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145488.1 | NP_663463.1 | 1 | 725 | ||
| GenBank mRNA | AK030565.1 | - | 1 | 708 | ||
| AK083898.1 | - | 1 | 713 | |||
| AK143173.1 | - | 1 | 713 | |||
| AK220525.1 | - | 1 | 706 | |||
| BC003424.1 | - | 1 | 725 | 2609884 | ||

Pex6
Genomic Map