siRNA Details
| Gene Symbol: Zfp959 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145490.4 | NP_663465.1 | 4 | 1415 | ||
| GenBank mRNA | BC011426.1 | - | 4 | 1388 | 3986622 | |
| BC138466.1 | - | 4 | 1363 | 8861486 | ||
| BC138467.1 | - | 4 | 1363 | 8861487 | ||

Zfp959
Genomic Map