siRNA Details
| Gene Symbol: Dpp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172624.3 | NP_766212.2 | 7 | 937 | ||
| GenBank mRNA | AK039652.1 | - | 6 | 1030 | ||
| AK050021.1 | - | 7 | 879 | |||
| AK050023.1 | - | 7 | 939 | |||
| AK078301.1 | - | 7 | 885 | |||
| AK131178.1 | - | 4 | 2800 | |||
| AK191928.1 | - | 2 | 47 | |||
| BC057631.1 | - | 7 | 936 | 4191886 | ||

Dpp9
Genomic Map