siRNA Details
| Gene Symbol: Dpp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172624.3 | NP_766212.2 | 3, 4 | 448 | ||
| GenBank mRNA | AK039652.1 | - | 3, 4 | 466 | ||
| AK050021.1 | - | 3, 4 | 390 | |||
| AK050023.1 | - | 3, 4 | 449 | |||
| AK078301.1 | - | 3, 4 | 396 | |||
| BC057631.1 | - | 3, 4 | 447 | 4191886 | ||

Dpp9
Genomic Map