siRNA Details
| Gene Symbol: 2410015M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153152.3 | NP_694792.1 | 2 | 259 | ||
| GenBank mRNA | AK075942.1 | - | 2 | 190 | ||
| AK075961.1 | - | 2 | 260 | |||
| BC056164.1 | - | 2 | 165 | 5713062 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2410015M20Rik| Gene Symbol: 2410015M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153152.3 | NP_694792.1 | 2 | 259 | ||
| GenBank mRNA | AK075942.1 | - | 2 | 190 | ||
| AK075961.1 | - | 2 | 260 | |||
| BC056164.1 | - | 2 | 165 | 5713062 | ||