siRNA Details
Gene Symbol: Vmn2r120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104591.1 | NP_001098061.1 | 3 | 321 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
