siRNA Details
| Gene Symbol: BC027072 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146082.3 | NP_666194.2 | 1 | 2216 | ||
| GenBank mRNA | BC046516.1 | - | 1 | 2216 | 5364819 | |
| BC060373.1 | - | 1 | 2225 | 6494066 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC027072| Gene Symbol: BC027072 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146082.3 | NP_666194.2 | 1 | 2216 | ||
| GenBank mRNA | BC046516.1 | - | 1 | 2216 | 5364819 | |
| BC060373.1 | - | 1 | 2225 | 6494066 | ||