siRNA Details
| Gene Symbol: Fez2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285940.1 | NP_001272869.1 | 8 | 1151 | ||
| NM_001285949.1 | NP_001272878.1 | 7 | 1070 | |||
| NM_199448.3 | NP_955519.1 | 7 | 1070 | |||
| GenBank mRNA | AK033547.1 | - | 7 | 1012 | ||
| AK150241.1 | - | 7 | 1036 | |||
| AK157221.1 | - | 7 | 1037 | |||
| AK159545.1 | - | 7 | 1023 | |||
| AK165595.1 | - | 7 | 1020 | |||
| AK218016.1 | - | 2 | 73 | |||
| AY382580.1 | - | 7 | 1021 | |||
| BC052366.1 | - | 8 | 1130 | |||
| BC096619.1 | - | 7 | 1015 | 6391451 | ||

Fez2
Genomic Map