siRNA Details
| Gene Symbol: Ttc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028639.3 | NP_082915.1 | 18 | 2417 | ||
| GenBank mRNA | AK040578.1 | - | 18 | 2420 | ||
| AK040610.1 | - | 18 | 2388 | |||
| AK050263.1 | - | 18 | 2404 | |||
| AK080284.1 | - | 18 | 2396 | |||
| AK153704.1 | - | 18 | 2399 | |||
| AK165244.1 | - | 11 | 1079 | |||
| BC023773.1 | - | 14 | 1375 | |||
| BC042512.1 | - | 18 | 2094 | 4223801 | ||
| BC049254.1 | - | 2 | 140 | |||
| BC057906.1 | - | 1 | 40 | |||

Ttc7
Genomic Map