siRNA Details
| Gene Symbol: Svil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153153.3 | NP_694793.1 | 6, 7 | 1945 | ||
| GenBank mRNA | AF317422.1 | - | 6, 7 | 1960 | ||
| AF317423.1 | - | 6, 7 | 1960 | |||
| AK041925.1 | - | 6, 7 | 1304 | |||
| BC115653.1 | - | 6, 7 | 1099 | |||
| BC115654.1 | - | 6, 7 | 1099 | |||
| BC128496.1 | - | 6, 7 | 1099 | |||
| BC128499.1 | - | 6, 7 | 1099 | |||
| BC152345.1 | - | 6, 7 | 1099 | |||
| BC152346.1 | - | 6, 7 | 1099 | |||
| HQ259291.1 | - | 3, 4 | 844 | |||

Svil
Genomic Map