siRNA Details
| Gene Symbol: Mib1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144860.2 | NP_659109.2 | 2 | 1051 | ||
| GenBank mRNA | AK139338.1 | - | 2 | 932 | ||
| AY147848.1 | - | 2 | 550 | |||
| AY149907.1 | - | 2 | 371 | |||
| AY245539.1 | - | 2 | 1051 | |||
| AY974091.1 | - | 2 | 344 | |||
| BC131662.1 | - | 3 | 517 | 40110696 | ||

Mib1
Genomic Map