siRNA Details
| Gene Symbol: Mib1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144860.2 | NP_659109.2 | 8 | 1847 | ||
| GenBank mRNA | AK053035.1 | - | 1 | 70 | ||
| AK080847.1 | - | 2 | 123 | |||
| AY147848.1 | - | 8 | 1346 | |||
| AY149907.1 | - | 8 | 1167 | |||
| AY245539.1 | - | 8 | 1847 | |||
| AY974091.1 | - | 8 | 1140 | |||
| BC131662.1 | - | 9 | 1313 | 40110696 | ||

Mib1
Genomic Map