siRNA Details
| Gene Symbol: Zfp521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145492.4 | NP_663467.1 | 4 | 3419 | ||
| GenBank mRNA | AK052655.1 | - | 4 | 3323 | ||
| AK077697.1 | - | 1 | 241 | |||
| AK081096.1 | - | 1 | 876 | |||
| AK131130.1 | - | 3 | 3504 | |||
| AY147406.1 | - | 4 | 3322 | |||
| BC021376.1 | - | 4 | 3235 | 5038671 | ||

Zfp521
Genomic Map