siRNA Details
| Gene Symbol: Klhl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081403.1 | NP_001074872.1 | 7 | 1721 | ||
| GenBank mRNA | AK018108.1 | - | 7 | 1436 | ||
| AK173163.1 | - | 7 | 1435 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl14| Gene Symbol: Klhl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081403.1 | NP_001074872.1 | 7 | 1721 | ||
| GenBank mRNA | AK018108.1 | - | 7 | 1436 | ||
| AK173163.1 | - | 7 | 1435 | |||