siRNA Details
| Gene Symbol: Fhod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289654.1 | NP_001276583.1 | 21 | 4643 | ||
| NM_001289655.1 | NP_001276584.1 | 22 | 5000 | |||
| NM_175276.4 | NP_780485.2 | 23 | 5096 | |||
| GenBank mRNA | AB078608.1 | - | 23 | 4274 | ||
| AB078609.1 | - | 21 | 3821 | |||
| AB100088.1 | - | 22 | 4178 | |||
| AK148245.1 | - | 21 | 3785 | |||
| AK220413.1 | - | 19 | 3260 | |||
| BC056371.1 | - | 13 | 2728 | |||
| BC060607.1 | - | 8 | 2052 | |||
| BC138046.1 | - | 21 | 3743 | 8861065 | ||

Fhod3
Genomic Map