siRNA Details
| Gene Symbol: Ammecr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242430.1 | NP_001229359.1 | 5 | 1310 | ||
| NM_153515.4 | NP_705735.1 | 5 | 1109 | |||
| GenBank mRNA | AK030009.1 | - | 5 | 1317 | ||
| AK054131.1 | - | 5 | 1108 | |||
| AK133018.1 | - | 5 | 1311 | |||
| AK151060.1 | - | 5 | 1117 | |||
| AK151257.1 | - | 5 | 1017 | |||
| BC012310.1 | - | 4 | 714 | |||
| BC034661.1 | - | 5 | 850 | 5137474 | ||
| BC056948.1 | - | 5 | 1053 | 6827318 | ||

Ammecr1l
Genomic Map