siRNA Details
| Gene Symbol: Reep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204914.1 | NP_001191843.1 | 6 | 715 | ||
| NM_144865.2 | NP_659114.2 | 6 | 721 | |||
| GenBank mRNA | AK147333.1 | - | 1 | 5415 | ||
| AK158694.1 | - | 1 | 295 | |||
| AY562230.1 | - | 6 | 554 | |||
| BC020184.1 | - | 6 | 652 | 4505679 | ||
| EU555417.1 | - | 6 | 548 | |||

Reep2
Genomic Map