siRNA Details
| Gene Symbol: Rbm27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172626.2 | NP_766214.2 | 17 | 2779 | ||
| GenBank mRNA | AK087759.1 | - | 4 | 359 | ||
| AK129330.1 | - | 12 | 1788 | |||
| AY461716.1 | - | 17 | 2754 | |||
| BC054080.1 | - | 13 | 2095 | 6390990 | ||
| BC059854.1 | - | 2 | 197 | |||

Rbm27
Genomic Map