siRNA Details
| Gene Symbol: Rbm27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042613.1 | - | 4 | 706 | ||
| AK129330.1 | - | 3 | 317 | |||
| BC054080.1 | - | 5 | 921 | 6390990 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm27| Gene Symbol: Rbm27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042613.1 | - | 4 | 706 | ||
| AK129330.1 | - | 3 | 317 | |||
| BC054080.1 | - | 5 | 921 | 6390990 | ||