siRNA Details
| Gene Symbol: Cep120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178686.3 | NP_848801.2 | 13 | 1877 | ||
| GenBank mRNA | AK038916.1 | - | 13 | 1939 | ||
| AK044808.1 | - | 12 | 2942 | |||
| AK047594.1 | - | 13 | 1922 | |||
| AK079319.1 | - | 13 | 1894 | |||
| AK089236.1 | - | 13 | 1884 | |||
| AK089242.1 | - | 13 | 1884 | |||
| BC046493.1 | - | 5 | 743 | |||
| BC053439.1 | - | 13 | 1877 | 6328829 | ||

Cep120
Genomic Map