siRNA Details
| Gene Symbol: Cep120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178686.3 | NP_848801.2 | 11 | 1575 | ||
| GenBank mRNA | AK038916.1 | - | 11 | 1637 | ||
| AK044808.1 | - | 10 | 2640 | |||
| AK047594.1 | - | 11 | 1620 | |||
| AK079319.1 | - | 11 | 1592 | |||
| AK089236.1 | - | 11 | 1582 | |||
| AK089242.1 | - | 11 | 1582 | |||
| BC046493.1 | - | 3 | 441 | |||
| BC053439.1 | - | 11 | 1575 | 6328829 | ||

Cep120
Genomic Map