siRNA Details
| Gene Symbol: Cplx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145493.1 | NP_663468.1 | 3 | 459 | ||
| GenBank mRNA | AK044262.1 | - | 3 | 467 | ||
| AY264291.1 | - | 3 | 348 | |||
| BC016082.1 | - | 3 | 459 | 4501240 | ||
| BC029011.1 | - | 3 | 407 | 5357047 | ||

Cplx4
Genomic Map