siRNA Details
| Gene Symbol: Cep76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081073.1 | NP_001074542.1 | 10 | 1515 | ||
| GenBank mRNA | AK035879.1 | - | 9 | 1295 | ||
| BC119216.1 | - | 10 | 1396 | 8733965 | ||
| BC119218.1 | - | 10 | 1396 | 8733967 | ||

Cep76
Genomic Map