siRNA Details
| Gene Symbol: Mapk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172632.2 | NP_766220.2 | 2 | 1473 | ||
| GenBank mRNA | AK079654.1 | - | 2 | 1451 | ||
| AK138531.1 | - | 1 | 1542 | |||
| AK162916.1 | - | 1 | 1542 | |||
| BC058942.1 | - | 1 | 876 | 6821544 | ||
| BC062911.1 | - | 1 | 876 | 6821544 | ||
| BC094564.1 | - | 2 | 1832 | |||

Mapk4
Genomic Map