siRNA Details
| Gene Symbol: St8sia5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013666.2 | NP_038694.2 | 4 | 596 | ||
| NM_153124.2 | NP_694764.2 | 3 | 488 | |||
| GenBank mRNA | AK013542.1 | - | 3 | 491 | ||
| AK078670.1 | - | 3 | 271 | |||
| AK162549.1 | - | 3 | 268 | |||
| BC034855.1 | - | 3 | 482 | 5146703 | ||
| BY733136.1 | - | 4 | 578 | |||
| X98014.1 | - | 4 | 459 | |||

St8sia5
Genomic Map