siRNA Details
| Gene Symbol: St8sia5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013666.2 | NP_038694.2 | 8 | 1122 | ||
| NM_153124.2 | NP_694764.2 | 7 | 1014 | |||
| GenBank mRNA | AK078670.1 | - | 7 | 797 | ||
| AK162549.1 | - | 7 | 794 | |||
| BC034855.1 | - | 7 | 1008 | 5146703 | ||
| X98014.1 | - | 8 | 985 | |||

St8sia5
Genomic Map