siRNA Details
| Gene Symbol: Haus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146089.2 | NP_666201.1 | 2 | 134 | ||
| GenBank mRNA | AK076912.1 | - | 2 | 114 | ||
| AK146923.1 | - | 2 | 131 | |||
| AK147599.1 | - | 2 | 141 | |||
| BC024400.1 | - | 2 | 134 | 5026575 | ||

Haus1
Genomic Map