siRNA Details
| Gene Symbol: Npas4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153553.4 | NP_705781.1 | 5 | 898 | ||
| GenBank mRNA | AB049835.1 | - | 5 | 793 | ||
| AK032607.1 | - | 4 | 1085 | |||
| AY730724.1 | - | 5 | 925 | |||
| BC129861.1 | - | 5 | 834 | 40110003 | ||

Npas4
Genomic Map