siRNA Details
| Gene Symbol: Kdm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001984.2 | NP_001001984.2 | 10, 11 | 1783 | ||
| GenBank mRNA | AK155866.1 | - | 10, 11 | 1788 | ||
| BC037117.1 | - | 3, 4 | 332 | |||
| BC076576.1 | - | 10, 11 | 1349 | 6840591 | ||

Kdm2a
Genomic Map